Assay Design



Gene Transcripts

Gene Symbol
CNGC13
Entrez Gene ID
826427
Gene Name
cyclic nucleotide-gated channel 13
Gene Aliases
AT4G01010, ATCNGC13, CYCLIC NUCLEOTIDE-GATED CHANNEL 13, F3I3.1, F3I3_1, cyclic nucleotide-gated channel 13
Chromosome Location
Chr.4: 434217 - 437605 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340295.1 NP_001319839.1 5 - 6 1386 155
NM_116329.3 NP_192010.2 4 - 5 1532 155

View Full Product Details