Assay Design



Gene Transcripts

Gene Symbol
GBF2
Entrez Gene ID
826505
Gene Name
G-box binding factor 2
Gene Aliases
AT4G01120, ATBZIP54, BASIC REGION/LEUCINE ZIPPER MOTIF 5, F2N1.12, F2N1_12, G-box binding factor 2
Chromosome Location
Chr.4: 481569 - 484524 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116342.3 NP_192021.1 4 - 5 397 79
GenBank mRNA AY062667.1 - 4 - 5 355 79
AY093316.1 - 3 - 4 214 79
X63895.1 - - 299 79

View Full Product Details