Assay Design



Gene Transcripts

Gene Symbol
MEE49
Entrez Gene ID
828136
Gene Name
Ribosomal RNA processing Brix domain protein
Gene Aliases
AT4G01560, F11O4.6, F11O4_6, maternal effect embryo arrest 49
Chromosome Location
Chr.4: 676982 - 679276 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116386.4 NP_567213.1 7 - 8 841 95
GenBank mRNA AY050341.1 - 7 - 8 773 95
AY086463.1 - 7 - 8 794 95
BT020609.1 - 7 - 8 729 95

View Full Product Details