Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001340355.1 | NP_001328437.1 | 10 - 11 | 1781 | 79 |
NM_001340356.1 | NP_001328436.1 | 10 - 11 | 1778 | 79 | |
NM_001340357.1 | NP_001328435.1 | 10 - 11 | 1778 | 79 | |
NM_116433.3 | NP_567221.1 | 10 - 11 | 1798 | 79 | |
GenBank mRNA | AF100163.1 | - | 10 - 11 | 1752 | 79 |
AY057477.1 | - | 10 - 11 | 1778 | 79 | |
AY090293.1 | - | 10 - 11 | 1685 | 79 |