Assay Design



Gene Transcripts

Gene Symbol
PRL
Entrez Gene ID
828153
Gene Name
Minichromosome maintenance (MCM2/3/5) family protein
Gene Aliases
AT4G02060, MCM7, PROLIFERA, T10M13.7, T10M13_7
Chromosome Location
Chr.4: 900636 - 905591 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203726.1 NP_001190655.1 5 - 6 319 74
NM_116437.3 NP_192115.1 3 - 4 269 74

View Full Product Details