Assay Design



Gene Transcripts

Gene Symbol
SYP42
Entrez Gene ID
827505
Gene Name
syntaxin of plants 42
Gene Aliases
AT4G02195, ATSYP42, ATTLG2B, TLG2B, syntaxin of plants 42
Chromosome Location
Chr.4: 969994 - 972342 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116452.4 NP_567223.1 2 - 3 622 76
GenBank mRNA AF154574.1 - 2 - 3 472 76
AK229319.1 - 2 - 3 584 76

View Full Product Details