Assay Design



Gene Transcripts

Gene Symbol
PARP2
Entrez Gene ID
828049
Gene Name
poly(ADP-ribose) polymerase
Gene Aliases
APP, AT4G02390, ATPARP1, PARP1, POLY(ADP-RIBOSE) POLYMERASE, POLY(ADP-RIBOSE) POLYMERASE 1, PP, T14P8.19, T14P8_19, poly(ADP-ribose) polymerase, poly(ADP-ribose) polymerase 2
Chromosome Location
Chr.4: 1049766 - 1054186 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340373.1 NP_001329308.1 11 - 12 1395 99
NM_001340374.1 NP_001329307.1 10 - 11 1462 99
NM_116472.4 NP_192148.2 11 - 12 1569 99

View Full Product Details