Assay Design



Gene Transcripts

Gene Symbol
APR1
Entrez Gene ID
825793
Gene Name
APS reductase 1
Gene Aliases
3'-PHOSPHOADENOSINE 5'-PHOSPHOSULFATE REDUCTASE, APR, APS REDUCTASE, APS reductase 1, AT4G04610, ATAPR1, F4H6.13, F4H6_13, PAPS REDUCTASE HOMOLOG 19, PRH19
Chromosome Location
Chr.4: 2324878 - 2327064 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116699.3 NP_192370.1 2 - 3 563 79
GenBank mRNA AF424582.1 - 2 - 3 435 79
BT002612.1 - 2 - 3 372 79

View Full Product Details