Assay Design



Gene Transcripts

Gene Symbol
MYB74
Entrez Gene ID
825855
Gene Name
myb domain protein 74
Gene Aliases
AT4G05100, AtMYB74, C17L7.20, C17L7_20, myb domain protein 74
Chromosome Location
Chr.4: 2618372 - 2619947 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116749.3 NP_192419.1 1 - 2 321 110
GenBank mRNA AF386932.1 - 1 - 2 239 110
BT001182.1 - 1 - 2 136 110

View Full Product Details