Assay Design



Gene Transcripts

Gene Symbol
SULTR3;5
Entrez Gene ID
832080
Gene Name
sulfate transporter 3;5
Gene Aliases
AT5G19600, T29J13.20, T29J13_20, sulfate transporter 3;5
Chromosome Location
Chr.5: 6612815 - 6617047 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121965.4 NP_568377.1 5 - 6 1046 123
GenBank mRNA AB061739.1 - 5 - 6 996 123
AK226785.1 - 5 - 6 1004 123

View Full Product Details