Assay Design



Gene Transcripts

Gene Symbol
IAA6
Entrez Gene ID
841717
Gene Name
indole-3-acetic acid 6
Gene Aliases
AT1G52830, F14G24.10, F14G24_10, SHORT HYPOCOTYL 1, SHY1, indole-3-acetic acid 6
Chromosome Location
Chr.1: 19672476 - 19673652 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104161.3 NP_175692.1 2 - 3 582 61
GenBank mRNA AB493505.1 - 2 - 3 489 61
AF336915.1 - 2 - 3 489 61

View Full Product Details