Assay Design
Gene Transcripts
- Gene Symbol
- ATMAP4K ALPHA1
- Entrez Gene ID
-
841750
- Gene Name
- Protein kinase superfamily protein
- Gene Aliases
-
AT1G53165, F8L10.20
- Chromosome Location
-
Chr.1: 19813790 - 19819564 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104195.3 |
NP_175724.2 |
13 - 14 |
1200 |
97 |
View Full Product Details