Assay Design



Gene Transcripts

Gene Symbol
ATMAP4K ALPHA1
Entrez Gene ID
841750
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT1G53165, F8L10.20
Chromosome Location
Chr.1: 19813790 - 19819564 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198280.1 NP_001185209.1 19 - 20 2094 90
NM_001333581.1 NP_001319212.1 19 - 20 2096 90
NM_104195.3 NP_175724.2 19 - 20 2084 90

View Full Product Details