Assay Design
Gene Transcripts
- Gene Symbol
- TTL1
- Entrez Gene ID
-
841764
- Gene Name
- tetratricopetide-repeat thioredoxin-like 1
- Gene Aliases
-
AT1G53300, F12M16.20, F12M16_20, tetratricopeptide-repeat thioredoxin-like 1, tetratricopetide-repeat thioredoxin-like 1
- Chromosome Location
-
Chr.1: 19879405 - 19882895 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104208.3 |
NP_175737.1 |
3 - 4 |
1629 |
107 |
GenBank mRNA |
BT006444.1 |
- |
3 - 4 |
1308 |
107 |
View Full Product Details