Assay Design



Gene Transcripts

Gene Symbol
NF-YA5
Entrez Gene ID
841856
Gene Name
nuclear factor Y, subunit A5
Gene Aliases
''nuclear factor Y, AT1G54160, F15I1.26, F15I1_26, NFYA5, NUCLEAR FACTOR Y A5, nuclear factor Y, subunit A5, subunit A5''
Chromosome Location
Chr.1: 20217336 - 20219452 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104294.3 NP_175818.1 3 - 4 926 72
GenBank mRNA AF386957.1 - 3 - 4 835 72
AY081445.1 - 2 - 3 636 72

View Full Product Details