Assay Design
Gene Transcripts
- Gene Symbol
- NHX5
- Entrez Gene ID
-
841878
- Gene Name
- sodium hydrogen exchanger 5
- Gene Aliases
-
AT1G54370, ATNHX5, F20D21.19, F20D21_19, NA+/H+ ANTIPORTER 5, sodium hydrogen exchanger 5
- Chromosome Location
-
Chr.1: 20293151 - 20297236 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104315.5 |
NP_175839.2 |
15 - 16 |
1156 |
98 |
GenBank mRNA |
AK226435.1 |
- |
15 - 16 |
1056 |
98 |
View Full Product Details