Assay Design



Gene Transcripts

Gene Symbol
NEK1
Entrez Gene ID
841893
Gene Name
NIMA-related serine/threonine kinase 1
Gene Aliases
AT1G54510, ATNEK1, F20D21.32, F20D21_32, NIMA-RELATED KINASE1, NIMA-related serine/threonine kinase 1
Chromosome Location
Chr.1: 20358319 - 20362624 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198295.1 NP_001185224.1 3 - 4 269 94
NM_001198296.1 NP_001185225.1 3 - 4 204 94
NM_001333653.1 NP_001319226.1 2 - 3 612 94
NM_104329.5 NP_175853.1 3 - 4 365 94

View Full Product Details