Assay Design



Gene Transcripts

Gene Symbol
LAS
Entrez Gene ID
842007
Gene Name
GRAS family transcription factor
Gene Aliases
AT1G55580, F20N2.1, Lateral Suppressor, SCARECROW-LIKE 18, SCL18
Chromosome Location
Chr.1: 20763844 - 20765823 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104434.4 NP_175954.1 1 - 1 1557 126
GenBank mRNA AY196482.1 - 1 - 1 1390 126
BT026519.1 - 1 - 1 1295 126

View Full Product Details