Assay Design



Gene Transcripts

Gene Symbol
GolS2
Entrez Gene ID
842114
Gene Name
galactinol synthase 2
Gene Aliases
AT1G56600, AtGolS2, F25P12.95, F25P12_95, galactinol synthase 2
Chromosome Location
Chr.1: 21207537 - 21209635 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104537.3 NP_176053.1 2 - 3 730 108
GenBank mRNA AB062849.1 - 2 - 3 726 108
AY050410.1 - 2 - 3 730 108
AY058238.1 - 2 - 3 647 108
KJ138960.1 - 2 - 3 647 108

View Full Product Details