Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001333852.1 | NP_001319271.1 | 8 - 9 | 1036 | 77 |
NM_001333853.1 | NP_001321061.1 | 8 - 9 | 1036 | 77 | |
NM_001333854.1 | NP_001321060.1 | 8 - 9 | 1036 | 77 | |
NM_104645.4 | NP_564743.1 | 8 - 9 | 1036 | 77 | |
NM_179496.2 | NP_849827.1 | 8 - 9 | 1021 | 77 | |
NM_179497.1 | NP_849828.1 | 8 - 9 | 934 | 77 | |
GenBank mRNA | AB028226.1 | - | 8 - 9 | 841 | 77 |
AK227804.1 | - | 8 - 9 | 918 | 77 | |
BT004825.1 | - | 8 - 9 | 729 | 77 |