Assay Design
Gene Transcripts
- Gene Symbol
- ULP1D
- Entrez Gene ID
-
842317
- Gene Name
- UB-like protease 1D
- Gene Aliases
-
AT1G60220, OTS1, OVERLY TOLERANT TO SALT 1, T13D8.11, T13D8_11, UB-like protease 1D
- Chromosome Location
-
Chr.1: 22208089 - 22212171 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104712.5 |
NP_176228.3 |
13 - 14 |
1900 |
65 |
GenBank mRNA |
DQ304543.1 |
- |
13 - 14 |
1657 |
65 |
View Full Product Details