Assay Design



Gene Transcripts

Gene Symbol
PANK1
Entrez Gene ID
842339
Gene Name
pantothenate kinase 1
Gene Aliases
AT1G60440, ATCOAA, ATPANK1, T13D8.31, T13D8_31, pantothenate kinase 1
Chromosome Location
Chr.1: 22266416 - 22269258 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333894.1 NP_001320517.1 5 - 6 885 115
NM_001333895.1 NP_001320518.1 5 - 6 649 115
NM_104731.5 NP_176247.2 6 - 7 781 115
GenBank mRNA AK229327.1 - 6 - 7 764 115
BT011005.1 - 6 - 7 631 115
BT015384.1 - 6 - 7 593 115

View Full Product Details