Assay Design



Gene Transcripts

Gene Symbol
ABC4
Entrez Gene ID
842354
Gene Name
UbiA prenyltransferase family protein
Gene Aliases
1, 4-DIHYDROXY-2-NAPTHOIC ACID PHYTYLTRANSFERASE, ABERRANT CHLOROPLAST DEVELOPMENT 4, AT1G60600, F8A5.36, F8A5_36
Chromosome Location
Chr.1: 22324437 - 22327359 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001124046.2 NP_001117518.1 7 - 8 1168 87
NM_104743.3 NP_176259.2 7 - 8 1160 87
GenBank mRNA AK227267.1 - 7 - 8 1091 87

View Full Product Details