Assay Design



Gene Transcripts

Gene Symbol
ACLA-2
Entrez Gene ID
842375
Gene Name
ATP-citrate lyase A-2
Gene Aliases
AT1G60810, ATP-citrate lyase A-2, F8A5.32, F8A5_32
Chromosome Location
Chr.1: 22388527 - 22391237 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333917.1 NP_001320572.1 11 - 12 1374 82
NM_104764.4 NP_176280.1 12 - 13 1326 82

View Full Product Details