Assay Design
Gene Transcripts
- Gene Symbol
- SCPL32
- Entrez Gene ID
-
842406
- Gene Name
- serine carboxypeptidase-like 32
- Gene Aliases
-
AT1G61130, F11P17.14, F11P17_14, serine carboxypeptidase-like 32
- Chromosome Location
-
Chr.1: 22528853 - 22531827 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104794.3 |
NP_176308.2 |
1 - 2 |
289 |
73 |
GenBank mRNA |
DQ056504.1 |
- |
1 - 2 |
246 |
73 |
View Full Product Details