Assay Design



Gene Transcripts

Gene Symbol
RPL3B
Entrez Gene ID
842454
Gene Name
R-protein L3 B
Gene Aliases
ARABIDOPSIS RIBOSOMAL PROTEIN 2, ARP2, AT1G61580, R-protein L3 B, RIBOSOMAL PROTEIN L3, T25B24.7, T25B24_7
Chromosome Location
Chr.1: 22720560 - 22723152 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104840.5 NP_176352.1 3 - 4 233 78
GenBank mRNA BT003869.1 - 3 - 4 237 78
BT005702.1 - 2 - 3 193 78

View Full Product Details