Assay Design



Gene Transcripts

Gene Symbol
TPS14
Entrez Gene ID
842465
Gene Name
terpene synthase 14
Gene Aliases
AT1G61680, ATTPS14, T13M11.3, T13M11_3, TERPENE SYNTHASE 14, terpene synthase 14
Chromosome Location
Chr.1: 22772315 - 22774720 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198357.1 NP_001185286.1 5 - 6 1207 104
NM_104850.2 NP_176361.2 5 - 6 1195 104

View Full Product Details