Assay Design



Gene Transcripts

Gene Symbol
MEE21
Entrez Gene ID
818021
Gene Name
PIN domain-like family protein
Gene Aliases
AT2G34570, T31E10.9, T31E10_9, maternal effect embryo arrest 21
Chromosome Location
Chr.2: 14562892 - 14564782 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129010.4 NP_181004.2 5 - 6 854 112
GenBank mRNA AY102549.1 - 5 - 6 854 112
AY954813.1 - 5 - 6 753 112

View Full Product Details