Assay Design



Gene Transcripts

Gene Symbol
TO1
Entrez Gene ID
818065
Gene Name
thioredoxin O1
Gene Aliases
AT2G35010, ATO1, F19I3.24, F19I3_24, thioredoxin O1
Chromosome Location
Chr.2: 14754234 - 14756323 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084537.1 NP_001078006.1 2 - 3 433 105
NM_129053.6 NP_181046.1 2 - 3 470 105
GenBank mRNA BT025773.1 - 2 - 3 306 105

View Full Product Details