Assay Design



Gene Transcripts

Gene Symbol
EYA
Entrez Gene ID
818099
Gene Name
eyes absent-like protein
Gene Aliases
AT2G35320, ATEYA, EYES ABSENT homolog, T4C15.1, T4C15_1
Chromosome Location
Chr.2: 14866832 - 14868881 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129084.3 NP_565803.1 3 - 4 533 64
GenBank mRNA AF410332.1 - 3 - 4 462 64
AY054555.1 - 3 - 4 487 64
AY064618.1 - 2 - 3 401 64

View Full Product Details