Assay Design



Gene Transcripts

Gene Symbol
MEE29
Entrez Gene ID
818101
Gene Name
helicase domain-containing protein
Gene Aliases
AT2G35340, T32F12.28, T32F12_28, maternal effect embryo arrest 29
Chromosome Location
Chr.2: 14872265 - 14879739 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336552.1 NP_001323587.1 7 - 8 476 144
NM_129086.4 NP_181077.3 11 - 12 1036 144

View Full Product Details