Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_105046.3 | NP_564815.1 | 7 - 8 | 1385 | 91 |
NM_202353.1 | NP_974082.1 | 7 - 8 | 1019 | 91 | |
GenBank mRNA | AY062807.1 | - | 7 - 8 | 972 | 91 |
BT008895.1 | - | 7 - 8 | 907 | 91 |