Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001334143.1 | NP_001320320.1 | 4 - 5 | 464 | 123 |
NM_105072.3 | NP_176582.2 | 4 - 5 | 464 | 123 | |
GenBank mRNA | AJ251990.1 | - | 4 - 5 | 453 | 123 |
DQ446392.1 | - | 4 - 5 | 430 | 123 |