Assay Design



Gene Transcripts

Gene Symbol
VHA-E3
Entrez Gene ID
842725
Gene Name
vacuolar H+-ATPase subunit E
Gene Aliases
AT1G64200, F22C12.4, F22C12_4, vacuolar H+-ATPase subunit E isoform 3
Chromosome Location
Chr.1: 23828284 - 23830100 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334149.1 NP_001320763.1 3 - 4 440 124
NM_001334150.1 NP_001320764.1 3 - 4 457 124
NM_105094.3 NP_176602.1 3 - 4 474 124

View Full Product Details