Assay Design



Gene Transcripts

Gene Symbol
SS2
Entrez Gene ID
820294
Gene Name
starch synthase 2
Gene Aliases
AT3G01180, AtSS2, T4P13.13, T4P13_13, starch synthase 2
Chromosome Location
Chr.3: 62245 - 66111 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_110984.3 NP_186767.1 2 - 3 1386 95
GenBank mRNA AY054467.1 - 2 - 3 1054 95
BT002555.1 - 2 - 3 953 95
KJ138848.1 - 2 - 3 953 95

View Full Product Details