Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_110984.3 | NP_186767.1 | 5 - 6 | 1639 | 88 |
GenBank mRNA | AY054467.1 | - | 5 - 6 | 1307 | 88 |
BT002555.1 | - | 5 - 6 | 1206 | 88 | |
KJ138848.1 | - | 5 - 6 | 1206 | 88 |