Assay Design



Gene Transcripts

Gene Symbol
HAK5
Entrez Gene ID
826973
Gene Name
high affinity K+ transporter 5
Gene Aliases
ARABIDOPSIS THALIANA HIGH AFFINITY K+ TRANSPORTER 5, AT4G13420, ATHAK5, T9E8.160, T9E8_160, high affinity K+ transporter 5
Chromosome Location
Chr.4: 7796782 - 7802251 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117416.3 NP_567404.1 7 - 8 1057 83
GenBank mRNA AF129478.1 - 7 - 8 989 83
BT010589.1 - 7 - 8 980 83

View Full Product Details