Assay Design
Gene Transcripts
- Gene Symbol
- HTA4
- Entrez Gene ID
-
826990
- Gene Name
- histone H2A 4
- Gene Aliases
-
AT4G13570, T6G15.120, T6G15_120, histone H2A 4
- Chromosome Location
-
Chr.4: 7884516 - 7885982 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117431.1 |
NP_193093.1 |
1 - 2 |
92 |
123 |
View Full Product Details