Assay Design
Gene Transcripts
- Gene Symbol
- PAP23
- Entrez Gene ID
-
827004
- Gene Name
- purple acid phosphatase 23
- Gene Aliases
-
AT4G13700, ATPAP23, F18A5.90, F18A5_90, purple acid phosphatase 23
- Chromosome Location
-
Chr.4: 7957072 - 7958919 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117444.3 |
NP_193106.3 |
5 - 6 |
1188 |
103 |
GenBank mRNA |
AY390530.1 |
- |
5 - 6 |
1188 |
103 |
View Full Product Details