Assay Design
Gene Transcripts
- Gene Symbol
- RLP48
- Entrez Gene ID
-
827022
- Gene Name
- receptor like protein 48
- Gene Aliases
-
AT4G13880, AtRLP48, F18A5.270, F18A5_270, receptor like protein 48
- Chromosome Location
-
Chr.4: 8025804 - 8028614 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117462.2 |
NP_193124.2 |
1 - 2 |
981 |
87 |
View Full Product Details