Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001340972.1 | NP_001328228.1 | 8 - 9 | 1371 | 87 |
NM_117564.4 | NP_193215.2 | 8 - 9 | 1456 | 87 | |
GenBank mRNA | AJ132843.1 | - | 8 - 9 | 1069 | 87 |
AY139995.1 | - | 8 - 9 | 1371 | 87 | |
BT008910.1 | - | 8 - 9 | 1048 | 87 |