Assay Design
Gene Transcripts
- Gene Symbol
- AO1
- Entrez Gene ID
-
827152
- Gene Name
- amine oxidase 1
- Gene Aliases
-
AT4G14940, ATAO1, DL3510W, FCAALL.145, amine oxidase 1
- Chromosome Location
-
Chr.4: 8541879 - 8544566 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117580.2 |
NP_193230.1 |
2 - 3 |
1244 |
119 |
View Full Product Details