Assay Design



Gene Transcripts

Gene Symbol
CYP97B3
Entrez Gene ID
827177
Gene Name
cytochrome P450, family 97, subfamily B, polypeptide 3
Gene Aliases
''cytochrome P450, AT4G15110, DL3600C, FCAALL.208, cytochrome P450, family 97, polypeptide 3, polypeptide 3'', subfamily B
Chromosome Location
Chr.4: 8629647 - 8633118 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117600.6 NP_193247.2 5 - 6 884 79
GenBank mRNA AY062675.1 - 5 - 6 792 79
BT002582.1 - 5 - 6 759 79

View Full Product Details