Assay Design



Gene Transcripts

Gene Symbol
CSLA01
Entrez Gene ID
827360
Gene Name
cellulose synthase-like A01
Gene Aliases
AT4G16590, ATCSLA01, ATCSLA1, CELLULOSE SYNTHASE-LIKE A1, DL4320W, FCAALL.402, cellulose synthase-like A01
Chromosome Location
Chr.4: 9345150 - 9349258 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341118.1 NP_001319964.1 8 - 9 1477 148

View Full Product Details