Assay Design



Gene Transcripts

Gene Symbol
MAP70-5
Entrez Gene ID
827434
Gene Name
microtubule-associated proteins 70-5
Gene Aliases
AT4G17220, ATMAP70-5, DL4645C, FCAALL.385, microtubule-associated proteins 70-5
Chromosome Location
Chr.4: 9656652 - 9659576 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117827.3 NP_193455.2 1 - 2 227 76
GenBank mRNA AK117336.1 - 1 - 2 141 76
AM086442.1 - 1 - 2 114 76
BT005901.1 - 1 - 2 56 76

View Full Product Details