Assay Design



Gene Transcripts

Gene Symbol
MAP70-5
Entrez Gene ID
827434
Gene Name
microtubule-associated proteins 70-5
Gene Aliases
AT4G17220, ATMAP70-5, DL4645C, FCAALL.385, microtubule-associated proteins 70-5
Chromosome Location
Chr.4: 9656652 - 9659576 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117827.3 NP_193455.2 3 - 4 384 74
GenBank mRNA AK117336.1 - 3 - 4 298 74
AM086442.1 - 3 - 4 271 74
BT005901.1 - 3 - 4 213 74

View Full Product Details