Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_117849.4 | NP_193476.1 | 2 - 3 | 823 | 87 |
GenBank mRNA | AJ441252.1 | - | 2 - 3 | 479 | 87 |
AK228587.1 | - | 2 - 3 | 661 | 87 | |
BT005756.1 | - | 2 - 3 | 661 | 87 | |
BT006112.1 | - | 2 - 3 | 479 | 87 | |
U09332.1 | - | 2 - 3 | 626 | 87 |