Assay Design



Gene Transcripts

Gene Symbol
IP5PII
Entrez Gene ID
827526
Gene Name
myo-inositol polyphosphate 5-phosphatase 2
Gene Aliases
4, 5)P3 5-PHOSPHATASE II, 5PTASE2, AT4G18010, AT5PTASE2, INOSITOL POLYPHOSPHATE 5-PHOSPHATASE II, INOSITOL(1, T6K21.190, T6K21_190, myo-inositol polyphosphate 5-phosphatase 2
Chromosome Location
Chr.4: 9990988 - 9994524 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341251.1 NP_001319979.1 3 - 4 572 68
NM_117911.4 NP_567547.1 3 - 4 663 68

View Full Product Details