Assay Design
Gene Transcripts
- Gene Symbol
- IP5PII
- Entrez Gene ID
-
827526
- Gene Name
- myo-inositol polyphosphate 5-phosphatase 2
- Gene Aliases
-
4, 5)P3 5-PHOSPHATASE II, 5PTASE2, AT4G18010, AT5PTASE2, INOSITOL POLYPHOSPHATE 5-PHOSPHATASE II, INOSITOL(1, T6K21.190, T6K21_190, myo-inositol polyphosphate 5-phosphatase 2
- Chromosome Location
-
Chr.4: 9990988 - 9994524 on Build TAIR10.1
View Full Product Details