Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001203432.1 | NP_001190361.1 | 3 - 4 | 772 | 81 |
NM_001343720.1 | NP_001332623.1 | 2 - 3 | 473 | 81 | |
NM_147894.4 | NP_680199.1 | 3 - 4 | 712 | 81 | |
GenBank mRNA | AK226614.1 | - | 3 - 4 | 717 | 81 |
AY302133.1 | - | 2 - 3 | 465 | 81 | |
BT022100.1 | - | 3 - 4 | 617 | 81 | |
BT026521.1 | - | 3 - 4 | 491 | 81 | |
KJ138673.1 | - | 3 - 4 | 491 | 81 |